Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INCENP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040694.1 | NP_001035784.1 | 18 | 2657 | ||
NM_020238.2 | NP_064623.2 | 17 | 2645 | |||
XM_006718533.2 | XP_006718596.1 | 18 | 2631 | |||
XM_011544995.2 | XP_011543297.1 | 18 | 2664 | |||
XM_011544996.2 | XP_011543298.1 | 18 | 2652 | |||
XM_011544997.2 | XP_011543299.1 | 17 | 2612 | |||
XM_011544998.2 | XP_011543300.1 | 17 | 2640 | |||
GenBank mRNA | AY714053.1 | - | 17 | 2455 | ||
BC098576.1 | - | 18 | 2657 | 30924748 |