Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077712.2 | NP_001071180.1 | 29 | 3496 | ||
NM_001290713.1 | NP_001277642.1 | 28 | 3222 | |||
NM_021465.4 | NP_067440.3 | 29 | 3356 | |||
GenBank mRNA | AJ002636.1 | - | 28 | 3198 | ||
AK034759.1 | - | 6 | 688 | |||
BC066041.1 | - | 29 | 3237 |