Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Star | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011485.4 | NP_035615.2 | 1 | 108 | ||
GenBank mRNA | AK014431.1 | - | 1 | 118 | ||
AK019725.2 | - | 1 | 148 | |||
AK054470.1 | - | 1 | 108 | |||
AK136327.1 | - | 1 | 118 | |||
BC060970.1 | - | 1 | 119 | |||
BC082283.1 | - | 1 | 119 | 30436512 | ||
L36062.1 | - | 1 | 124 |