Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Star | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011485.4 | NP_035615.2 | 3 | 258 | ||
GenBank mRNA | AK014431.1 | - | 3 | 267 | ||
AK019725.2 | - | 3 | 298 | |||
AK054470.1 | - | 3 | 258 | |||
AK136327.1 | - | 3 | 268 | |||
BC060970.1 | - | 3 | 269 | |||
BC082283.1 | - | 3 | 269 | 30436512 | ||
L36062.1 | - | 3 | 274 |