Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109905.2 | NP_001103375.1 | 3 | 472 | ||
NM_001109906.2 | NP_001103376.1 | 3 | 472 | |||
NM_001291160.1 | NP_001278089.1 | 3 | 472 | |||
NM_011490.4 | NP_035620.1 | 3 | 472 | |||
GenBank mRNA | AF061942.1 | - | 3 | 472 | ||
AF395842.1 | - | 2 | 193 | |||
AK005002.2 | - | 3 | 413 | |||
AK139532.1 | - | 3 | 419 | |||
AK145826.1 | - | 3 | 439 | |||
AK151835.1 | - | 3 | 278 | |||
AK152269.1 | - | 3 | 284 | |||
AK152744.1 | - | 3 | 278 | |||
AK159602.1 | - | 3 | 389 | |||
AK159781.1 | - | 3 | 442 | |||
AK160979.1 | - | 3 | 413 | |||
AK167637.1 | - | 3 | 413 | |||
AY391773.1 | - | 2 | 205 | |||
BC012959.1 | - | 3 | 436 | 4188394 | ||
BC082277.1 | - | 3 | 409 |