Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015801.1 | - | 4 | 556 | ||
AF129870.1 | - | 4 | 934 | |||
AF145287.1 | - | 4 | 538 | |||
AF151711.1 | - | 4 | 538 | |||
AK004499.1 | - | 2 | 217 | |||
AK147802.1 | - | 4 | 597 | |||
AK171909.1 | - | 5 | 1332 | |||
AK172385.1 | - | 4 | 1321 | |||
AK172528.1 | - | 4 | 1318 | |||
BC052379.1 | - | 4 | 599 | 5688695 | ||
EU730638.1 | - | 4 | 517 |