Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297434.1 | NP_001284363.1 | 4 | 400 | ||
NM_005540.2 | NP_005531.2 | 10 | 907 | |||
XM_006710621.3 | XP_006710684.2 | 6 | 670 | |||
XM_011541392.2 | XP_011539694.1 | 4 | 346 | |||
XM_011541393.2 | XP_011539695.1 | 4 | 335 | |||
XM_017001206.1 | XP_016856695.1 | 10 | 944 | |||
XM_017001207.1 | XP_016856696.1 | 7 | 647 | |||
XM_017001208.1 | XP_016856697.1 | 7 | 776 | |||
XM_017001209.1 | XP_016856698.1 | 4 | 326 | |||
GenBank mRNA | AK022846.1 | - | 4 | 358 | ||
AK295062.1 | - | 2 | 133 | |||
AK310824.1 | - | 10 | 897 | |||
BC042529.2 | - | 3 | 282 | |||
BC058932.1 | - | 10 | 871 | 5497202 | ||
DC398675.1 | - | 4 | 400 | |||
M74161.1 | - | 10 | 901 |