Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198977.1 | NP_001185906.1 | 2 | 351 | ||
NM_011518.2 | NP_035648.2 | 2 | 355 | |||
GenBank mRNA | AK036736.1 | - | 2 | 279 | ||
AK049207.1 | - | 3 | 449 | |||
AK136363.1 | - | 2 | 354 | |||
AK138741.1 | - | 2 | 286 | |||
AK143564.1 | - | 2 | 384 | |||
BB629720.1 | - | 2 | 277 | |||
BC065121.1 | - | 2 | 287 | 30533300 | ||
U25685.1 | - | 1 | 139 | |||
U36776.1 | - | 2 | 358 | |||
Z49877.1 | - | 2 | 616 |