Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002195.1 | NP_002186.1 | 2 | 435 | ||
GenBank mRNA | AK291543.1 | - | 2 | 435 | ||
BC026254.1 | - | 2 | 452 | 4770395 | ||
L34838.1 | - | 2 | 435 |