Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011539.3 | NP_035669.3 | 9 | 1140 | ||
GenBank mRNA | AK048544.1 | - | 9 | 1122 | ||
AK085414.1 | - | 6 | 892 | |||
AK139479.1 | - | 9 | 1103 | |||
BC029014.1 | - | 9 | 1142 | 5007629 | ||
L18868.1 | - | 9 | 1011 |