Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291269.1 | NP_001278198.1 | 2, 3 | 357 | ||
NM_009330.3 | NP_033356.2 | 2, 3 | 726 | |||
GenBank mRNA | AB008174.1 | - | 2, 3 | 546 | ||
AB008176.1 | - | 2, 3 | 546 | |||
AB052659.1 | - | 2, 3 | 546 | |||
AK004837.2 | - | 2, 3 | 727 | |||
BC025189.1 | - | 2, 3 | 357 | 5044299 | ||
X55842.1 | - | 2, 3 | 546 |