Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289603.1 | NP_001276532.1 | 14, 15 | 1548 | ||
NM_033476.3 | NP_258437.1 | 14, 15 | 1554 | |||
GenBank mRNA | AK013652.1 | - | 1, 2 | 68 | ||
AK141248.1 | - | 8, 9 | 894 | |||
AK146503.1 | - | 14, 15 | 1555 | |||
AK158682.1 | - | 14, 15 | 1474 | |||
BC022131.1 | - | 14, 15 | 1521 | 5098336 | ||
M84809.1 | - | 14, 15 | 1493 |