Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009342.2 | NP_033368.1 | 3, 4 | 286 | ||
GenBank mRNA | AK004520.1 | - | 40 | |||
AK133788.1 | - | 3, 4 | 216 | |||
AK153971.1 | - | 3, 4 | 221 | |||
BC043018.1 | - | 3, 4 | 286 | 5684255 | ||
BC089472.1 | - | 3, 4 | 230 | 6703917 | ||
M25825.1 | - | 198 |
Gene Symbol: Dynlt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166629.2 | NP_001160101.1 | 3, 4 | 260 | ||
GenBank mRNA | BC087868.1 | - | 3, 4 | 260 | 6740486 |
Gene Symbol: Dynlt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166630.1 | NP_001160102.1 | 3, 4 | 217 | ||
GenBank mRNA | AK168478.1 | - | 3, 4 | 218 | ||
EF110999.1 | - | 189 |
Gene Symbol: Dynlt1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166627.1 | NP_001160099.1 | 3, 4 | 325 | ||
NM_001199948.1 | NP_001186877.1 | 3, 4 | 325 | |||
GenBank mRNA | AK005569.1 | - | 3, 4 | 205 | ||
BY099269.1 | - | 3, 4 | 218 | |||
BY250333.1 | - | 3, 4 | 325 |