Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011550.1 | - | 3 | 441 | ||
AK076365.1 | - | 1 | 1368 | |||
AK133474.1 | - | 3 | 409 | |||
AK139088.1 | - | 3 | 647 | |||
AK144793.1 | - | 3 | 625 | |||
AK155268.1 | - | 1 | 1386 | |||
BC015077.1 | - | 3 | 324 | |||
BC145595.1 | - | 2 | 415 | |||
BC145596.1 | - | 3 | 330 | |||
U81490.1 | - | 3 | 381 |