Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011550.1 | - | 10, 11 | 1163 | ||
AK076365.1 | - | 8, 9 | 2079 | |||
AK133474.1 | - | 10, 11 | 1131 | |||
AK139088.1 | - | 10, 11 | 1369 | |||
AK144793.1 | - | 10, 11 | 1347 | |||
AK155268.1 | - | 7, 8 | 2208 | |||
AK193704.1 | - | 3, 4 | 233 | |||
BC015077.1 | - | 10, 11 | 1045 | |||
BC145595.1 | - | 9, 10 | 1137 | |||
BC145596.1 | - | 10, 11 | 1052 | |||
U81490.1 | - | 10, 11 | 1103 |