Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011564.1 | NP_035694.1 | 1 | 1111 | ||
GenBank mRNA | BC111528.2 | - | 2 | 1048 | 40045188 |