Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207176.3 | NP_997059.1 | 6 | 1146 | ||
GenBank mRNA | AK132249.1 | - | 6 | 1104 | ||
AK145609.1 | - | 6 | 1095 | |||
AK167101.1 | - | 6 | 1095 | |||
AK167274.1 | - | 6 | 1103 | |||
BC003808.1 | - | 8 | 1386 | 3589585 | ||
BC010465.1 | - | 6 | 1102 | 3585670 | ||
BC051103.1 | - | 6 | 1078 | |||
CT010319.1 | - | 5 | 981 | |||
X78989.1 | - | 5 | 975 |