Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011574.2 | NP_035704.2 | 8 | 1056 | ||
GenBank mRNA | AK144455.1 | - | 8 | 1041 | ||
AK146263.1 | - | 8 | 1032 | |||
AK150862.1 | - | 8 | 1031 | |||
AK168470.1 | - | 8 | 1006 | |||
AK173318.1 | - | 7 | 997 | |||
BC027399.1 | - | 4 | 555 | 4948207 | ||
BC068113.1 | - | 8 | 1045 | 5707865 |