Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011591.4 | NP_035721.1 | 3 | 187 | ||
GenBank mRNA | AF106621.1 | - | 3 | 141 | ||
AK003204.1 | - | 3 | 160 | |||
AK009164.1 | - | 3 | 189 | |||
AK011830.1 | - | 3 | 188 | |||
AK049584.1 | - | 3 | 188 | |||
AK150836.1 | - | 3 | 190 | |||
AK151396.1 | - | 3 | 195 | |||
AK153097.1 | - | 3 | 188 | |||
BC008275.1 | - | 3 | 169 | 2811777 |