Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083927.1 | NP_001077396.1 | 20 | 3363 | ||
NM_001083928.1 | NP_001077397.1 | 20 | 3309 | |||
NM_009389.2 | NP_033415.2 | 20 | 3333 | |||
GenBank mRNA | AK031322.1 | - | 18 | 2140 | ||
AK122524.1 | - | 20 | 2817 | |||
AK147635.1 | - | 11 | 7461 | |||
AK158785.1 | - | 19 | 2644 | |||
AK189910.1 | - | 3 | 286 | |||
AK198921.1 | - | 4 | 320 | |||
BC006672.1 | - | 9 | 1285 | |||
BC056465.1 | - | 20 | 2344 | 5709252 | ||
X73360.1 | - | 19 | 2516 |