Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019634.2 | NP_062608.2 | 4 | 410 | ||
GenBank mRNA | AF052492.1 | - | 4 | 358 | ||
AK004776.1 | - | 4 | 370 | |||
AK147306.1 | - | 4 | 372 | |||
AK147404.1 | - | 4 | 374 | |||
BC026516.1 | - | 3 | 209 | 4911317 | ||
BC050153.1 | - | 4 | 358 | |||
BC058603.1 | - | 4 | 366 | 6529011 | ||
D26483.1 | - | 4 | 358 |