Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009398.2 | NP_033424.1 | 6 | 808 | ||
GenBank mRNA | AK139985.1 | - | 6 | 818 | ||
BC021155.1 | - | 6 | 787 | 4208738 | ||
U83903.1 | - | 6 | 808 |