Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 14 | 1804 | ||
NR_073103.1 | 14 | 1804 | ||||
NR_073104.1 | 14 | 1804 | ||||
NR_073105.1 | 14 | 1804 | ||||
NR_073106.1 | 14 | 1804 | ||||
NR_073107.1 | 13 | 1683 | ||||
GenBank mRNA | AK307952.1 | - | 14 | 1793 | ||
AU126174.1 | - | 4 | 484 | |||
BC143505.1 | - | 14 | 1690 | |||
BC143508.1 | - | 14 | 1690 | |||
BC143509.1 | - | 14 | 1690 | |||
BC143511.1 | - | 14 | 1690 | |||
BC143512.1 | - | 13 | 1569 | |||
X17033.1 | - | 14 | 1709 |