Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011635.1 | NP_035765.1 | 1 | 402 | ||
GenBank mRNA | AK146737.1 | - | 1 | 217 | ||
BC051644.1 | - | 1 | 195 | 30022193 | ||
M36387.1 | - | 1 | 402 |