Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011638.4 | NP_035768.1 | 18 | 2132 | ||
GenBank mRNA | AK088961.1 | - | 18 | 2046 | ||
BC054522.1 | - | 18 | 2017 | 6406097 | ||
BC067403.1 | - | 4 | 748 | |||
X57349.1 | - | 17 | 1968 |