Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009443.3 | NP_033469.1 | 2, 3 | 1246 | ||
GenBank mRNA | AK008520.1 | - | 1, 2 | 15 | ||
AK041302.1 | - | 2, 3 | 996 | |||
AK076586.1 | - | 2, 3 | 1247 | |||
AK133559.1 | - | 2, 3 | 994 | |||
AK161242.1 | - | 2, 3 | 979 | |||
AK162574.1 | - | 2, 3 | 996 | |||
BC009143.1 | - | 2, 3 | 981 | 3157708 | ||
BC141218.1 | - | 2, 3 | 991 | 9055830 | ||
BC145542.1 | - | 2, 3 | 991 | 9054105 | ||
CT010269.1 | - | 2, 3 | 968 | |||
D50031.1 | - | 2, 3 | 994 |