Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009446.2 | NP_033472.1 | 4 | 721 | ||
GenBank mRNA | AK132775.1 | - | 4 | 721 | ||
BC050770.1 | - | 4 | 707 | 6774775 | ||
BC089547.1 | - | 4 | 707 | 6771690 | ||
M13442.1 | - | 4 | 709 |
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009449.3 | NP_033475.1 | 4 | 773 | ||
GenBank mRNA | BC050769.1 | - | 4 | 697 | 6771187 | |
M13443.1 | - | 4 | 688 |