Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002208.4 | NP_002199.3 | 24, 25 | 3021 | ||
XM_011523823.2 | XP_011522125.1 | 24, 25 | 3522 | |||
XM_011523825.2 | XP_011522127.1 | 24, 25 | 3520 | |||
XM_011523827.2 | XP_011522129.1 | 23, 24 | 3352 | |||
XM_011523828.2 | XP_011522130.1 | 22, 23 | 3278 | |||
XM_017024586.1 | XP_016880075.1 | 24, 25 | 4829 | |||
GenBank mRNA | BC113436.1 | - | 24, 25 | 3011 | 8322488 | |
BC117207.1 | - | 24, 25 | 3011 | 40125758 | ||
L25851.2 | - | 24, 25 | 3047 |