Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009450.2 | NP_033476.1 | 4 | 1476 | ||
GenBank mRNA | AK134545.1 | - | 4 | 1475 | ||
BC003475.1 | - | 1 | 956 | |||
BC055441.1 | - | 4 | 1466 | 4016867 | ||
CB522913.1 | - | 1 | 290 |