Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020557.4 | NP_065582.3 | 1 | 771 | ||
GenBank mRNA | AK004595.1 | - | 1 | 606 | ||
AK150512.1 | - | 1 | 596 | |||
AK150543.1 | - | 1 | 598 | |||
AK150725.1 | - | 1 | 596 | |||
AK150885.1 | - | 1 | 598 | |||
AK153429.1 | - | 1 | 600 | |||
AK153469.1 | - | 1 | 603 | |||
BC027329.1 | - | 1 | 670 | 3982036 | ||
BC048910.1 | - | 1 | 782 | |||
BC057565.1 | - | 1 | 584 | 6401867 |