Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033962.1 | NP_001029134.1 | 4 | 2039 | ||
NM_011668.2 | NP_035798.2 | 6 | 2226 | |||
NM_173010.3 | NP_766598.1 | 4 | 2041 | |||
GenBank mRNA | AK018028.1 | - | 1 | 17 | ||
AK029133.1 | - | 4 | 2042 | |||
AK083795.1 | - | 4 | 1971 | |||
AK209747.1 | - | 2 | 359 | |||
AK220549.1 | - | 4 | 1939 | |||
BC131658.1 | - | 4 | 2027 | 40110636 | ||
BC131659.1 | - | 4 | 2027 | 40110637 | ||
U82122.1 | - | 2 | 1476 | |||
U96636.1 | - | 6 | 1773 |