Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093771.2 | NP_001087240.1 | 10 | 1213 | ||
NM_001261445.1 | NP_001248374.1 | 8 | 1635 | |||
NM_001261446.1 | NP_001248375.1 | 7 | 1190 | |||
NM_003330.3 | NP_003321.3 | 8 | 1550 | |||
NM_182729.2 | NP_877393.1 | 8 | 1313 | |||
NM_182742.2 | NP_877419.1 | 7 | 1440 | |||
NM_182743.2 | NP_877420.1 | 7 | 1203 | |||
GenBank mRNA | AF077367.1 | - | 7 | 867 | ||
AF208018.1 | - | 6 | 763 | |||
AJ001050.1 | - | 6 | 776 | |||
AK128515.1 | - | 7 | 1098 | |||
AK226071.1 | - | 8 | 1021 | |||
AK293322.1 | - | 7 | 850 | |||
AK295067.1 | - | 8 | 949 | |||
AK296495.1 | - | 7 | 840 | |||
AK304241.1 | - | 10 | 1196 | |||
AK308881.1 | - | 8 | 1244 | |||
AK312262.1 | - | 8 | 1212 | |||
AY057105.1 | - | 10 | 1189 | |||
BC018122.1 | - | 8 | 926 | 3883490 | ||
BT019640.1 | - | 6 | 739 | |||
CR536506.1 | - | 6 | 739 | |||
D88687.1 | - | 8 | 991 | |||
S79851.1 | - | 8 | 1178 | |||
X91247.1 | - | 8 | 1178 |