Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083319.1 | NP_001076788.1 | 4 | 703 | ||
NM_013699.2 | NP_038727.1 | 4 | 703 | |||
GenBank mRNA | AB221581.1 | - | 3 | 292 | ||
AK143282.1 | - | 4 | 704 | |||
AY182062.1 | - | 3 | 292 | |||
BC034749.1 | - | 4 | 392 | |||
BC139828.1 | - | 3 | 320 | 40142981 | ||
BC141082.1 | - | 3 | 382 | 9055694 | ||
U20086.1 | - | 3 | 294 |