Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009461.2 | NP_033487.2 | 18 | 2172 | ||
GenBank mRNA | AF061555.1 | - | 18 | 2173 | ||
AK089616.1 | - | 18 | 2072 | |||
AK132284.1 | - | 18 | 2086 | |||
AK166885.1 | - | 18 | 2070 | |||
BC090969.1 | - | 18 | 2060 | |||
BC100461.1 | - | 18 | 2079 |