Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013701.3 | NP_038729.1 | 1 | 279 | ||
GenBank mRNA | AK002629.1 | - | 1 | 280 | ||
AY227195.1 | - | 1 | 289 | |||
BC138676.1 | - | 1 | 278 | 8861697 | ||
BC145969.1 | - | 1 | 278 | 40131272 | ||
D87866.1 | - | 1 | 253 |