Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040691.1 | NP_001035781.1 | 2 | 374 | ||
NM_011677.2 | NP_035807.2 | 1 | 346 | |||
GenBank mRNA | AK146178.1 | - | 2 | 374 | ||
AK168709.1 | - | 1 | 296 | |||
AK168785.1 | - | 1 | 296 | |||
BC011039.1 | - | 2 | 431 | 4009947 | ||
BC052853.1 | - | 1 | 290 | 30058519 | ||
U55041.1 | - | 1 | 300 | |||
X99018.1 | - | 1 | 302 | |||
Y08975.1 | - | 2 | 374 |