Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011680.2 | NP_035810.1 | 4, 5 | 483 | ||
GenBank mRNA | AB221562.1 | - | 4, 5 | 442 | ||
AK146871.1 | - | 4, 5 | 430 | |||
AK158906.1 | - | 2, 3 | 259 | |||
BC082995.1 | - | 4, 5 | 483 | 6440967 | ||
U01662.1 | - | 4, 5 | 499 |