Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009497.3 | NP_033523.1 | 2 | 174 | ||
GenBank mRNA | AF007168.1 | - | 1 | 76 | ||
AK090178.1 | - | 2 | 193 | |||
BC039745.1 | - | 2 | 155 | 3156993 | ||
BC055105.1 | - | 2 | 174 | 6596132 | ||
U60150.1 | - | 2 | 136 |