Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011692.2 | NP_035822.2 | 6 | 581 | ||
GenBank mRNA | AK002282.1 | - | 6 | 551 | ||
AK008847.1 | - | 6 | 551 | |||
AK077923.1 | - | 6 | 566 | |||
AK151527.1 | - | 6 | 551 | |||
AK153069.1 | - | 6 | 550 | |||
AK159284.1 | - | 6 | 551 | |||
AK167064.1 | - | 6 | 551 | |||
AK169185.1 | - | 6 | 551 | |||
BC132169.1 | - | 6 | 524 | |||
BC132171.1 | - | 6 | 524 | |||
DV066998.1 | - | 6 | 614 |