Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161420.1 | NP_001154892.1 | 10 | 1932 | ||
NM_013703.2 | NP_038731.2 | 10 | 1932 | |||
GenBank mRNA | AK076172.1 | - | 10 | 1937 | ||
AK136838.1 | - | 10 | 1752 | |||
BC013622.1 | - | 10 | 1368 | 3968213 | ||
L33417.1 | - | 10 | 1356 | |||
U06670.1 | - | 10 | 1518 |