Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018765.3 | NP_061235.2 | 2, 3 | 415 | ||
GenBank mRNA | AF071184.1 | - | 2, 3 | 267 | ||
AK019160.1 | - | 2, 3 | 149 | |||
AK135215.1 | - | 2, 3 | 415 | |||
AK159522.1 | - | 2, 3 | 667 | |||
AK160559.1 | - | 2, 3 | 152 | |||
AK161496.1 | - | 2, 3 | 109 | |||
AK162009.1 | - | 2, 3 | 157 |