Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011719.4 | NP_035849.3 | 2 | 219 | ||
GenBank mRNA | AB073819.1 | - | 2 | 182 | ||
AF469004.1 | - | 2 | 167 | |||
AK052708.1 | - | 2 | 220 | |||
BC110482.1 | - | 2 | 211 | 40048674 |