Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023653.5 | NP_076142.3 | 3 | 661 | ||
GenBank mRNA | AK012093.1 | - | 3 | 577 | ||
AK045120.1 | - | 3 | 573 | |||
AK167642.1 | - | 3 | 577 | |||
BC026373.1 | - | 3 | 556 | 4162686 |