Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271757.1 | NP_001258686.1 | 2, 3 | 312 | ||
NM_001271758.1 | NP_001258687.1 | 2, 3 | 255 | |||
NM_009525.3 | NP_033551.2 | 2, 3 | 509 | |||
GenBank mRNA | AK013218.1 | - | 2, 3 | 480 | ||
AK049558.1 | - | 2, 3 | 279 | |||
BC010775.1 | - | 2, 3 | 255 | 4211437 | ||
BC051406.1 | - | 2, 3 | 440 | 6438917 | ||
M89799.1 | - | 1, 2 | 124 |