Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009531.2 | NP_033557.2 | 5 | 633 | ||
GenBank mRNA | AB071144.1 | - | 5 | 570 | ||
AK004713.1 | - | 5 | 573 | |||
AK028595.1 | - | 5 | 571 | |||
AK166981.1 | - | 5 | 568 | |||
AK170975.1 | - | 5 | 565 | |||
BC052725.1 | - | 5 | 595 | 6812198 | ||
U27398.1 | - | 5 | 541 |