Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009531.2 | NP_033557.2 | 2 | 332 | ||
GenBank mRNA | AB071144.1 | - | 2 | 269 | ||
AK004713.1 | - | 2 | 272 | |||
AK028595.1 | - | 2 | 270 | |||
AK166981.1 | - | 2 | 267 | |||
AK170975.1 | - | 2 | 264 | |||
AK191690.1 | - | 1 | 152 | |||
BC052725.1 | - | 2 | 294 | 6812198 | ||
BY238377.1 | - | 2 | 333 | |||
U40005.1 | - | 1 | 71 |