Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205132.1 | NP_001192061.1 | 4 | 610 | ||
NM_001205133.1 | NP_001192062.1 | 4 | 610 | |||
NM_009535.3 | NP_033561.1 | 4 | 1072 | |||
GenBank mRNA | AK035612.1 | - | 4 | 1059 | ||
AK052478.1 | - | 4 | 562 | |||
AK167420.1 | - | 4 | 747 | |||
X67677.1 | - | 4 | 999 |