Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194333.3 | NP_919314.2 | 12 | 1808 | ||
GenBank mRNA | BC094893.1 | - | 12 | 1811 | 30017276 | |
BC117790.1 | - | 12 | 1795 | 40092152 | ||
BC117791.1 | - | 12 | 1795 | 40092153 | ||
U25739.1 | - | 12 | 1820 |