Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011738.2 | NP_035868.1 | 2 | 440 | ||
GenBank mRNA | AF077002.1 | - | 2 | 365 | ||
AK077596.1 | - | 2 | 303 | |||
AK149224.1 | - | 2 | 410 | |||
AK166922.1 | - | 2 | 239 | |||
AK168520.1 | - | 2 | 302 | |||
AK169035.1 | - | 2 | 291 | |||
AK169189.1 | - | 2 | 239 | |||
AK187410.1 | - | 2 | 140 | |||
AK217599.1 | - | 1 | 51 | |||
BC008187.1 | - | 2 | 229 | 3589806 | ||
BC061497.1 | - | 2 | 384 | 4949445 | ||
CB183421.1 | - | 2 | 440 | |||
D87661.1 | - | 2 | 372 | |||
U57311.1 | - | 2 | 188 |