Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324236.1 | NP_001311165.1 | 2 | 347 | ||
NM_001324237.1 | NP_001311166.1 | 3 | 252 | |||
NM_001324238.1 | NP_001311167.1 | 2 | 255 | |||
NM_001324240.1 | NP_001311169.1 | 3 | 344 | |||
NM_033453.3 | NP_258412.1 | 3 | 344 | |||
NM_181493.3 | NP_852470.1 | 3 | 293 | |||
NR_052001.2 | 2 | 265 | ||||
XM_006723564.3 | XP_006723627.1 | 3 | 627 | |||
XM_011529234.2 | XP_011527536.1 | 3 | 627 | |||
GenBank mRNA | AB062127.1 | - | 3 | 213 | ||
AF026816.2 | - | 3 | 203 | |||
AF063607.1 | - | 3 | 228 | |||
AF219116.1 | - | 3 | 256 | |||
AF334680.1 | - | 2 | 136 | |||
AK309165.1 | - | 2 | 206 | |||
AV694278.1 | - | 2 | 137 | |||
BC010138.2 | - | 3 | 188 | 4124624 | ||
BE275997.1 | - | 2 | 222 | |||
BM560812.1 | - | 344 | ||||
BM703982.1 | - | 2 | 164 | |||
EF199841.1 | - | 3 | 191 | |||
EF213026.1 | - | 3 | 140 | |||
KU178148.1 | - | 3 | 152 |